Labconco™ Screen Purifier™ Starter Kits
For use with Purifier™ Logic™ Biosafety Cabinet
Manufacturer: Labconco™ LCN3406101
Description
- Includes washable/reusable, metal screen-type prefilter, hardware and bracket
- For applications needing control of large debris, animal hair and dander
Specifications
Prefilters and postfiters | |
4 ft. Purifier logic and biosafety cabinet | |
2.3kg | |
Screen Prefilter Kit |
Needing control of large debris, animal hair and dander | |
Washable/reusable, metal screen-type prefilter, hardware and bracket | |
5 lb. |