Labconco™ Replacement Prefilter for Protector Downdraft Powder Station
Replacement Prefilter for use in Protector Downdraft Powder Stations.
Supplier: Labconco™ 3768903
Description
Replacement Prefilter for use in Protector Downdraft Powder Stations.
Replacement Prefilter for use with Labconco Protector Downdraft Powder Station forensic enclosures. Made of pleated paper. MERV 8 rating.Specifications
Prefilter | |
0.8 in. | |
0.5 Kg | |
17.8 in. | |
37.5 cm | |
Prefilter | |
Particulate |
Protector Downdraft Powder Station | |
1 lb. | |
14.8 in. | |
1.9 cm | |
45.1 cm | |
Paper |