Please login to your online account to display your discounted pricing
Labconco™ Replacement Filters for Benchtop Fume Adsorbers
Manufacturer: labconco™  LCN6961302
 View more versions of this product
Catalog No. 10-090-128
  • / Each
Description & Specifications
Specifications
Product Type Filter
Filter Type Carbon
For Use With (Equipment) Benchtop fume adsorber
For Use With (Application) Ammonia-amine
Weight (English) 11 lb.
Weight (Metric) 5kg