Please login to your online account to display your discounted pricing
Labconco™ Replacement Beaker for Fat/Oil Extraction
Specifically designed for use with Labconco Goldfisch Fat Extractor
$129.32 - $300.09
Specifications
For Use With (Application) Goldfisch fat extraction
For Use With (Equipment) Fat/oil extractor
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
09-680-10
labconco™
LCN3506200
Reclaiming Tube Pack of 6 Each for $129.32
09-680-14
labconco™
LCN3502200
Heating Element Each for $177.92
09-680-5
labconco™
LCN3505300
Replacement Beaker Pack of 6 Each for $300.09
09-680-8
labconco™
LCN3506100
Sample Tube Pack of 6 Each for $213.55
LCN3502200
labconco™
3502200
Heating Element Each for $202.00
Description & Specifications
Specifications
For Use With (Application) Goldfisch fat extraction
For Use With (Equipment) Fat/oil extractor
  • Capacity: 100mL
  • Thick-walled
  • Borosilicate glass
  • With flanged and ground top to facilitate connection to condenser