Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Horizontal Clean Bench, 3 ft. Width


Protect worker from particulate contamination with the Labconco Purifier™ Horizontal Clean Bench.

Manufacturer: labconco™  LCN3889700

 View more versions of this product

Supporting base (sold separately).

 Alert

Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.

Catalog No. 13-118-100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Electrical Requirements 115V, 60Hz, 6A, Domestic
Plug Type North America
Depth (English) 34 in.
Material Stainless Steel
Width (Metric) Exterior 96.5cm
Filter Efficiency 99.99%
Height (Metric) Exterior 122.9cm
Width (English) Exterior 38 in.
Filter Type HEPA
Ports Utility
Net Weight (Metric) 165kg
For Use With (Application) Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
Depth (Metric) 86.4cm
Depth (English) Interior 21 in.
Certifications/Compliance CAN/CSA C22.2, ETL, UL, CE
Depth (Metric) Interior 53cm
Lighting Fluorescent
Height (English) Exterior 48.4 in.
Net Weight (English) 364 lb.