Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Horizontal Clean Bench, 3 ft. Width


Protect worker from particulate contamination with the Labconco Purifier™ Horizontal Clean Bench.

Manufacturer: labconco™  LCN3889720

 View more versions of this product

Supporting base (sold separately).

 Alert

Units do not protect user and should not be used in conjunction with biohazardous material, toxins or radionuclides.

Catalog No. 13-118-110

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Plug Type North America
Electrical Requirements 115V, 60Hz, 6A, Domestic
Net Weight (English) 364 lb.
Net Weight (Metric) 165kg
Lighting Fluorescent, UV
Ports Utility
Filter Efficiency 99.99%
Filter Type HEPA
Depth (English) Interior 21 in.
Certifications/Compliance CAN/CSA C22.2, ETL, UL, CE
Material Stainless Steel
Depth (Metric) Interior 53cm
Width (English) Exterior 38 in.
Width (Metric) Exterior 96.5cm
Height (English) Exterior 48.4 in.
Height (Metric) Exterior 122.9cm
Depth (English) 34 in.
Depth (Metric) 86.4cm
For Use With (Application) Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation