Please login to your online account to display your discounted pricing

Labconco™ Purifier™ HEPA-Filtered Safety Enclosure, Without Guardian Monitor


Ensure personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ HEPA-Filtered Safety Enclosure.

Manufacturer: labconco™  LCN3980200

 View more versions of this product

Catalog No. 10-304-122

  • / Each
 

Description & Specifications

Specifications

Weight (English) 125 lb.
Volume 110 to 155 cfm
Filter Type HEPA
Housing Material Epoxy Coated Aluminum and Steel
Type Laminar Flow Cabinet
Amperage 3A
Voltage 115V
Height (English) 37.08 in.
Lighting Fluorescent
Hertz 60Hz
Weight (Metric) 57kg
Sash Opening (Metric) 23.97cm
Width (English) 23.75 in.
Width (Metric) 60.32cm
Electrical Requirements 115V 60Hz
Sash Opening (Metric) 23.97cm