Please login to your online account to display your discounted pricing

Labconco™ Purifier™ HEPA-Filtered Safety Enclosures with Guardian Monitor


Labconco Purifier™ HEPA-Filtered Safety Enclosures with Guardian Monitor provide personnel and environmental biohazard protection.

Manufacturer: labconco™  LCN3980302

 View more versions of this product

Catalog No. 10-304-127

  • / Each
 

Description & Specifications

Specifications

Sash Opening (Metric) 23.97cm
Width (English) 35.75 in.
Width (Metric) 90.8cm
Amperage 3A
Height (English) 37.08 in.
Type Laminar flow safety cabinet
Hertz 60Hz
Housing Material Epoxy Coated Aluminum and Steel
Lighting Fluorescent
Voltage 115V
Volume 165 to 230 cfm
Weight (English) 160 lb.
Weight (Metric) 72.57kg
Electrical Requirements 115V 60Hz
Filter Type HEPA
Sash Opening (Metric) 23.97cm