missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Base Stands for Horizontal Clean Bench, 5 ft.

Add a Labconco Purifier™ Base Stand to yourHorizontal Clean Bench.

Manufacturer:  Labconco™ LCN3255000

 View more versions of this product

Catalog No. 32-550-00



Description

Description

These base stands have telescoping legs so that they may be adjusted to eight height positions (height selection and adjustment must be completed before the enclosure is installed atop the stand). The stands are available with fixed feet or casters for mobility. • Support any Labconco Purifier Clean Bench or Enclosure. • Support loads up to 1000 pounds. • ADA-compliant. • Durable 1.75 in. tubular, powder-coated steel frame. Models 3' wide and larger have rear crossbar supports. • Telescoping legs are adjustable to eight height positions in 1 in. increments from 27.5 in. to 34.5 in.. • Available with fixed leveling feet or 5 in. diameter, toe-locking, non-marking polyurethane casters with bearings for quiet operation.
Specifications

Specifications

29 in.
Epoxy Coated Steel
64.5 in.
28 in.
130 lb.
Utility
73.66cm
5 ft. Telescoping Base Stand with fixed feet
163.83cm
71.12cm
58.9kg
SDS
Documents