missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet

Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.

Manufacturer:  Labconco™ LCN304410300

 View more versions of this product

Catalog No. 10-304-200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
 
  • Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
  • Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes
Specifications

Specifications

32.7 in.
115V, 60Hz, 12A, Domestic
245.1cm
Fluorescent
Safety Cabinet
5 years
326.59kg
138cm
96.5 in.
Two GFCI duplex
Class II Safety Cabinet
83.1cm
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Base stand and Bag-In/Bag-Out Exhaust Filter
North America
10 in.
720 lb.
54.2 in.
NSF International-Listed, ETL, Modified ASHRAE 110
200w
25.4cm
480 CFM @ 0.2 in. s.p.
Documents