missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet

Labconco Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinet features ergonomic Inclination™ Technology and MyLogic™ Operating System.

Specifications

Product Type Safety Cabinet
Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110
For Use With (Application) Antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Sash Opening (Metric) 25.4cm
Width (Metric) 137.6cm
View More Specs

Products 8
Catalog Number Mfr. No. Plug Type Includes Electrical Requirements Height (Metric) Price Quantity  
Catalog Number Mfr. No. Plug Type Includes Electrical Requirements Height (Metric) Price Quantity  
30-441-0000
View Documents
Labconco™
LCN304410000
North America N/A 115V, 60Hz, 12A, Domestic 163cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-1000
View Documents
Labconco™
LCN304411000
North America Service fixture, Vacu-Pass Portal 115V, 60Hz, 12A, Domestic 163cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-0100
View Documents
Labconco™
LCN304410100
North America Base stand 115V, 60Hz, 12A, Domestic 233cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-1100
View Documents
Labconco™
LCN304411100
North America Service fixture, Vacu-Pass Portal, Base Stand 115V, 60Hz, 12A, Domestic 233cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-0010
View Documents
Labconco™
LCN304410010
North America N/A 230V, 50/60Hz, 6A, Domestic 163cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-1010
View Documents
Labconco™
LCN304411010
North America Service fixture, Vacu-Pass Portal 230V, 50/60Hz, 6A, Domestic 163cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-0110
View Documents
Labconco™
LCN304410110
North America Base stand 230V, 50/60Hz, 6A, Domestic 233cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-441-1110
View Documents
Labconco™
LCN304411110
North America Service fixture, Vacu-Pass Portal, Base Stand 230V, 50/60Hz, 6A, Domestic 233cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
 

  • Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
  • Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes

C1 designation is pending NSF approval

Specifications

Specifications

Safety Cabinet
Antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
137.6cm
Fluorescent
NSF International-Listed, ETL, Modified ASHRAE 110
25.4cm
81.2cm
SDS