missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Protector™ Stainless-Steel Perchloric Acid Hoods, Deep

Type 316 stainless-steel interior withstands reaction from perchloric acid

Manufacturer:  Labconco™ LCN130410001

 View more versions of this product

Catalog No. 13-118-206

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Requires : Remote blowers, ductwork, base cabinets, adjustable height base stands

Specifications Description & Specifications

Specifications

59 in.
48 in.
50/60Hz
Epoxy Coated Steel
115V, 50/60Hz
Fume Hood
149.9cm
121.9cm
115V
Fluorescent
725cfm at 0.25 in. H2O at 100fpm