missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Protector™ Fiberglass Filtered Glove Boxes
Features inlet and outlet HEPA filters, 99.99% efficient on particulates
Supplier: Labconco™ 5065010
Requires : Supporting base
Description
- One-piece molded fiberglass liner
- Clear acrylic inner and outer transfer chamber doors
- Two glove ports
- 0.3μm particulate containment <20 ng/m3
- Built-in blower with speed control capable of airflow from 15 CFM to 85 CFM
- 0.05 ppm average tracer gas levels (surpasses ASHRAE 110 standards)
- Dry powder-coated, 18 and 20 gauge steel exterior panel
- Bag-In/Bag-Out filter disposal system
- Space-saving inner and outer transfer chamber doors pivoting upward equipped with quick-latches
- Two 8 in. ID, epoxy-coated aluminum, double-grooved glove ports mounted 17 in. apart with neoprene gaskets
- Two 25-watt fluorescent lights that exceed 120 footcandles
- LCD display for real time monitoring of volume dilution rates in cubic feet per minute, air changes/minute, and main chamber pressure (inches of water)
- Interior left-hand side wall-mounted pass-through plug
- Interior right-hand side wall-mounted electrical receptacle
- With pair of neoprene gloves, 0.015 in. thick, 30 in. long
- Large viewing area, 36W x 27.7 in. H with 0.375 in. thick, laminated safety glass
- Dimensions: 63.9W x 31.0D x 45.7 in. H (162.3 x 78.7 x 116.1cm); Interior working width: 35.5 in.
Work with hazardous materials
Certifications
Exceeds ISO Class 4 conditions and achieves ISO Class 3 conditions, CE marked (International models)
Compliance
ETL listing, UL 61010-1 and CAN/CSA C22.2 No. 1010.1 conformity (North America models)
Chemical Identifiers
| Glove Box | |
| Fluorescent (2 x 25 W) | |
| 10 A | |
| 78.7 cm | |
| 45.7 in. | |
| 50/60 Hz | |
| 500 lb. | |
| 63.9 in. | |
| 162.3 cm |
| HEPA | |
| Fiberglass | |
| 31 in. | |
| LCD | |
| 116.1 cm | |
| 2 | |
| 227 kg | |
| 35 in. | |
| 88.9 cm |
Specifications
| Glove Box | |
| Fluorescent (2 x 25 W) | |
| 10 A | |
| 31 in. | |
| LCD | |
| 116.1 cm | |
| North America Plug | |
| 100/115 V | |
| 227 kg | |
| 35 in. | |
| 88.9 cm |
| HEPA | |
| Fiberglass | |
| UL, ETL, CAN/CSA | |
| 78.7 cm | |
| 45.7 in. | |
| 50/60 Hz | |
| 2 | |
| 500 lb. | |
| 63.9 in. | |
| 162.3 cm |