Please login to your online account to display your discounted pricing

Labconco™ Protector™ Evidence Drying Cabinets with Washdown

Spray nozzle allows the interior to be washed down for easy clean up after the cabinet has been properly disinfected

Specifications

Cabinet Type Evidence drying lockable cabinet
Depth (English) Cabinet 28 in.
Depth (English) Shelf 19 in.
Depth (Metric) Cabinet 71.1cm
Depth (Metric) Shelf 48.26cm
View More Specs
Products
Catalog Number Mfr. No. Amperage Electrical Requirements Plug Type Price Quantity    

09-682-793

 
labconco corp
LCN3400010
3A 115V 60Hz Domestic CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Cabinet Type Evidence drying lockable cabinet
Depth (English) Cabinet 28 in.
Depth (English) Shelf 19 in.
Depth (Metric) Cabinet 71.1cm
Depth (Metric) Shelf 48.26cm
Material Stainless Steel lined
Product Type Evidence Drying Cabinet
Door Type Safety glass door with magnetic seal
Filter Type HEPA and Carbon filter
Height (English) Cabinet 79 in.
Height (English) Chamber 12 in.
Height (Metric) Cabinet 200.7cm
Height (Metric) Chamber 30.48cm
Height (Metric) Storage Area 30.48cm
Includes Washdown system with pump
Weight (English) 450 lb.
Weight (Metric) 204kg
Width (English) Cabinet 36 in.
Width (English) Shelf 8 in.
Width (English) Storage Area 36 in.
Width (Metric) Cabinet 91.44cm
Width (Metric) Shelf 20.3cm
Width (Metric) Storage Area 91.44cm
With Casters Yes

Key Features

  • Provides a secure ventilated area to dry evidence such as clothing and bedding
  • HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors

Additional Features

  • 2 in. (5.1cm) OD drain with 7.9 gal./min. (30L/min.) drain pump and washdown spray nozzle with 6 ft. (1.8m) coiled hose for additional decontamination
  • Type 304 brushed stainless steel interior, stainless steel rod and three stainless steel collapsible/removable wire mesh shelves, 8W × 19 in.D (20 × 48cm)
  • Safety glass door with magnetic seal that accommodates a user-supplied padlock or optional tamper-evident tag
  • Epoxy-coated steel exterior
  • Storage area, 17.5W × 26.5D × 12 in.H (44.5 × 67.3 × 30.5cm) with door
  • 2 in. (5.1cm) diameter locking casters
  • Inlet and exhaust roughing prefilters with MERV 11 rating; exhaust HEPA filter, 99.99% efficient on particles 0.3μm; activated organic vapor carbon filter, 7.5 lb. (3.4kg) porous coconut shell media
  • Built-in blower with solid state speed control
  • Red filter system alarm light to alert operator to low airflow conditions
  • 96 in. (2.4m) power cord and plug

ETL listed (Mfr. Nos. 3400010, 3400011), CE marked (Mfr. Nos. 3400012, 3400013, 3400014)