Please login to your online account to display your discounted pricing

Labconco™ Protector™ Evidence Drying Cabinet: 5 ft.W


Provide a compact, space-saving method of securely drying evidence in your lab with the Labconco Protector Evidence Drying Cabinet. This durable, stainless steel-lined cabinet provides a secure ventilated area to dry evidence such as clothing and bedding and its HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors.

Manufacturer: labconco™  LCN3405004

 View more versions of this product

Catalog No. 09-682-752

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Width (English) Storage Area 36 in.
Material Stainless Steel lined
Width (English) 60 in.
Depth (Metric) Cabinet 71.1cm
Depth (English) Cabinet 28 in.
Electrical Requirements 230V 50/60Hz
Depth (English) Shelf 19 in.
Width (Metric) Shelf 20.3cm
With Casters Yes
Plug Type China/Australia plug
Height (English) Chamber 12 in.
Height (Metric) Cabinet 200.7cm
Depth (English) 26.5 in.
Width (Metric) Cabinet 152.4cm
Depth (Metric) 67.31cm
Amperage 3A
Height (Metric) 200.7cm
Height (Metric) Storage Area 30.48cm
Height (English) Storage Area 12 in.
Voltage 220V
Door Type Safety glass door with magnetic seal
Product Type Evidence Drying Cabinet
Depth (Metric) Shelf 48.26cm
Width (English) Cabinet 60 in.
Height (English) Cabinet 79 in.
Includes UV light
Width (English) Shelf 8 in.
Height (English) 79 in.
Width (Metric) 152.4cm
Hertz 50/60Hz
Height (Metric) Chamber 30.48cm
Filter Type HEPA and Carbon filter
Width (Metric) Storage Area 91.44cm
Weight (Metric) 306kg
Options Available UV Light
Weight (English) 675 lb.
Cabinet Type Evidence drying lockable cabinet

A built-in blower provides airflow through the cabinet interior to speed drying. Air passes through the door inlet prefilter, circulates up through the interior, passes through the exhaust prefilter, HEPA
and carbon filters and is exhausted back to the room through the top of the cabinet.

  • Durable, stainless steel-lined construction
  • HEPA and carbon-filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors
  • Ultraviolet light provides secondary disinfection of interior between cases to prevent cross contamination, ensuring integrity of samples for DNA testing
  • Lockable cabinet maintains the chain of custody
  • UV security lock limits access to the ultraviolet light switch
  • Safety glass door with magnetic seal maximizes visibility into the cabinet while withstanding exposure to ultraviolet light
  • Inlet roughing prefilter traps dust and other large particles in the air entering the cabinet
  • Storage area below cabinet houses drain bucket and holds miscellaneous cleaning supplies
  • Leak-tight brushed stainless-steel interior cleans easily
  • Three-position switch activates fan or ultraviolet light (only when UV security lock key is in ON position)
  • Spacious interior measures 26.5D x 32.25W x 56 in.H (81.9 x 67.3 x 142.2cm)
  • Stainless-steel hanging rod accommodates clothing, bedding and other tall or bulky items; three wire mesh shelves
  • Exhaust roughing prefilter traps particulates in the exhaust air to prolong the life of the HEPA filter
  • HEPA filter, 99.99% efficient on particles 0.3 micron, traps airborne particulates including biohazardous aerosols, as air is exhausted from cabinet interior
  • Large 14-quart plastic bucket, located in the storage area below the cabinet, collects liquids from the drain in the cabinet's floor to aid clean up