Please login to your online account to display your discounted pricing

Labconco™ Protector™ Echo™ Filtered Benchtop Fume Hoods: 4 ft. Width

Adaptable fully-featured fume hood with Neutrodine™ filtration technology

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 121.9cm
View More Specs
Products
Catalog Number Mfr. No. Amperage Includes Voltage Weight (Metric) Price Quantity    

13-118-301

 
labconco™
LCN180410000
10Amp Integral blower 100-115V 240kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-302

 
labconco™
LCN180410001
10Amp Integral blower, 2 service fixtures 100-115V 244.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-303

 
labconco™
LCN180410002
10Amp Integral blower, 2 service fixtures, 1 electrical duplex 100-115V 244.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-304

 
labconco™
LCN181410000
10Amp Side windows, integral blower 100-115V 254kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-305

 
labconco™
LCN181410001
10Amp Side windows, integral blower, 2 service fixtures 100-115V 258.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-306

 
labconco™
LCN181410002
10Amp Side windows, integral blower, 2 service fixtures, 1 electrical duplex 100-115V 258.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-307

 
labconco™
LCN182410000
10Amp Back and side windows, integral blower 100-115V 258.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-308

 
labconco™
LCN182410001
10Amp Back and side windows, integral blower, 2 service fixtures 100-115V 263.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-309

 
labconco™
LCN182410002
10Amp Back and side windows, integral blower, 2 service fixtures, 1 electrical duplex 100-115V 263.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-310

 
labconco™
LCN180410020
5Amp Integral blower 208-230V 240.4kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-311

 
labconco™
LCN180410021
5Amp Integral blower, 2 service fixtures 208-230V 244.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-313

 
labconco™
LCN181410020
5Amp Side windows, integral blower 208-230V 254kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-314

 
labconco™
LCN181410021
5Amp Side windows, integral blower; 2 service fixtures 208-230V 258.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-316

 
labconco™
LCN182410020
5Amp Back and side windows, integral blower 208-230V 258.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-317

 
labconco™
LCN182410021
5Amp Back and side windows, integral blower; 2 service fixtures 208-230V 263.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 121.9cm

AFNOR NF-X, ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, SEFA 8 (Cabinet Surface Finish), SEFA 9, UL 61010