missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Protector™ Acid Digestion Hoods: 6ft. Width, 115V

PVC hood with bypass airflow design draws away harmful acid fumes

Manufacturer:  Labconco™ LCN141610000

 View more versions of this product

Catalog No. 13-118-190

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Requires : Remote blowers, ductwork, base cabinets, adjustable height base stands

Specifications Description & Specifications

Specifications

37.7 in.
59 in.
72 in.
28 in. high
50/60Hz
1180cfm at 0.38 in. H2O at 100fpm
254kg
ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, CFR 29, NFPA 45, SEFA 1, SEFA 8
T8 fluorescent
1180cfm at 0.38 in. H2O at 100fpm
95.8cm
149.9cm
182.9cm
71.1cm high
115V
560 lb.
10Amp
None
115V, 50/60Hz
Benchtop