Labconco™ Pressure Gauge Kits for Precise™ and HEPA-Filtered Glove Boxes
Accessories for use with Precise Glove Boxes and HEPA-Filtered Glove Boxes, XPert Weigh Boxes
$630.76 - $1,051.28
Specifications
For Use With (Application) Monitors pressures between 0 to 29 in. of mercury
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Weight (English) Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Type Weight (English) Weight (Metric) Price Quantity  
16-108-419 Labconco™
LCN5237900
Transfer chamber pressure gauge kit 3 lb. 1.4kg Each for $1,051.28
16-108-418 Labconco™
LCN5237800
Main chamber pressure gauge kit 3 lb. 1.4kg Each for $1,051.28
16-108-420 Labconco™
LCN5238100
Transfer chamber pressure gauge kit 2 lb. 0.9kg Each for $630.76
Specifications Description & Specifications
Specifications
Monitors pressures between 0 to 29 in. of mercury