Please login to your online account to display your discounted pricing
Labconco™ Prefilters for Paramount™ Filtered Enclosures
Replace prefilters when changing filter cells
$60.68 - $366.52
Specifications
For Use With (Equipment) 72 in. W Paramount™ filtered enclosures
Weight (Metric) 1.4kg
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
16-304-73
labconco™
LCN6911000
Roughing Prefilter Pack of 2 Each for $60.68
16-304-74
labconco™
LCN6912000
Electrostatic Prefilter Pack of 2 Each for $366.52
Description & Specifications
Specifications
For Use With (Equipment) 72 in. W Paramount™ filtered enclosures
Weight (Metric) 1.4kg
  • Choose roughing prefilters, as originally supplied, or electrostatic prefilters