Labconco™ Precise™ Glove Box Base Stands
For use with Precise™ glove boxes
Manufacturer:  Labconco™ LCN5235500
 View more versions of this product
Catalog No. 10-358-109
Specifications
Description
Base Stand; For Precise Glove Boxes; Black laminate work surface; Lower shelf for accessories; 31D x 60W x 33 to 40 in.H (79 x 152 x 84 to 102cm); Mobile with casters
  • Height-adjustable from 83.8 to 101.6cm (33 to 40 in.)
  • Permanently mounted black laminate work surface
  • Lower shelf holds accessory equipment
  • With casters or leveling feet
Description & Specifications
Specifications
40 in.
152.4cm
Mobile
General lab use
101.6 to 83.8cm
225 lb.
101.6cm
60 in.
Precise™ glove box, Protector™ glove box, XPert™ weigh box
40 to 33 in.
5 in. casters
102kg