Labconco™ Paramount™ Ductless Enclosures: 4ft. Wide
Conveniently added fume elimination to your lab. The Labconco Paramount™ Ductless Enclosure requires no outside ducting.
Manufacturer:  Labconco™ LCN6963401
 View more versions of this product
Catalog No. 10-030-209
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Requires : Filters, work surface, base, identical or no more than two different types of filters may be used in one enclosure: 4 ft. models require 6 filters
Specifications Description & Specifications
Specifications
32 in.
29 in.
200kg
122cm
230 to 380 cfm
440 lb.
30.5cm
115V 60Hz
57 to 67dBA
12 in.
74cm
135cm
Acid-Sulfur, Ammonia-Amine, Formaldehyde, HEPA, Mixed Bed, Organic Vapor, Radioisotope
Enclosure
48 in.
5A
53 in.
ANSI Z9.5-2003, CAN/CSA C22.2 No. 1010.1, UL 3101-1/61010-1, ASHRAE 110-95, SEFA 1-2005
115V
60Hz