Labconco™ Lyph-Lock™ Flask Adapters
Flask Adapters: For connecting Lyph-Lock flasks to valve ports on drying chamber or manifold
Manufacturer:  Labconco™ LCN7568200
 View more versions of this product
Catalog No. 10-269-69B
Specifications Description & Specifications
Specifications
For connecting lyph-lock flasks to valve ports on drying chamber or manifold
90° bend
Flask Adapter
0.5 lb.
Lyph lock flasks
Borosilicate Glass
19/38 to 0.75 in. port
0.2kg