Please login to your online account to display your discounted pricing

Labconco™ LabSolutions™ Powder Detergent


Removes grease, blood, agar, protein digestates, and other contaminants

Manufacturer: labconco™  LCN4422100

 View more versions of this product

Catalog No. 04-335

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For use in Glassware washers, removes range of contaminants including Protein digestates, Agar, Blood and Grease
pH Range 12.5
Size 27.5 lb.