Please login to your online account to display your discounted pricing

Labconco™ Guardian™ Airflow Monitor


Senses and alerts the operator to low airflow conditions

Manufacturer: labconco™  LCN9413301

 View more versions of this product

Catalog No. 10-369-205

  • / Each
 

Description & Specifications

Specifications

Product Type Guardian Airflow Monitor
Model Guardian Airflow Monitor, 220V
Weight (Metric) 2.7kg
Weight (English) 6 lb.
Alarm Range 30-250 fpm
Electrical Requirements 208-230V, 50/60Hz
For Use With (Equipment) Protector Premier Laboratory Hoods, Protector XL Laboratory Hoods, Protector XStream Laboratory Hood