Labconco™ G4 Replacement Prefilters
For use with Purifier™ Logic™ Biosafety Cabinet
Manufacturer: Labconco™ LCN3406200
Description
- Includes pleated disposable filter with Merv 8 rating
- Replacement filter cells for Purifier Logic™ and Cell Logic™ Disposable Prefilter Kit
Specifications
Prefilters and postfiters | |
Pleated disposable filter with Merv 8 rating | |
2 lb. |
3 ft. Purifier logic and biosafety cabinet | |
0.9kg | |
G4 Replacement Prefilter |