missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ FreeZone™ Console Freeze Dry System, 12 Liters

Designed for moderate aqueous sample loads

Specifications

Capacity 3.17 gal., 12L
Voltage 230V
For Use With (Application) For moderate aqueous sample loads
Material Stainless Steel, Epoxy-coated Steel Exterior
Style Console
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
10-271-77 Labconco™
LCN7754041
Ptfe coated collector N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
16-208-5015 Labconco™
LCN7754046
Mini chamber, purge valve N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
16-208-5018 Labconco™
LCN7759042
Purge valve, stoppering tray dryer N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Removes maximum of 8 liters of water in 24 hours
  • Upright stainless-steel collector coil
  • CFC-free refrigeration system to cool collector to -50°C (-58°F)
  • Corrosion-resistant glacier white epoxy-coated steel and brushed stainless steel exterior with blue accents
  • LCD for display of system operating parameters, set up parameters and alarm messages
  • Automatic start-up switch for collector cool down and vacuum pull down with manual override switches
  • Retractable, 9 in. collector drain line
  • Power switch and rear-mounted electrical receptacle (for vacuum pump connection)
  • Features 0.75 in. (1.9cm) OD vacuum connection, three feet of 0.75 in. (1.9cm) ID vacuum hose and two clamps

  • Purge valve with switch available
  • Stoppering tray dryer with three shelves for holding serum bottles or vials
  • Microprocessor-controlled temperature ramp and hold programming from -40°C to +40°C
  • Memory stores five programs and repetition of identical protocols
  • Control panel with operating switches, stoppering control knob, vacuum release valve control knob
  • Red LED alarm indicator
  • Hot gas defrost and switch

Compliance

CE compliant

Description & Specifications

Specifications

3.17 gal., 12L
For moderate aqueous sample loads
Console
230V
Stainless Steel, Epoxy-coated Steel Exterior