Please login to your online account to display your discounted pricing

Labconco™ FreeZone™ Bulk Tray Dryers

For use with 6, 12, or 18L FreeZone Freeze-Dry Systems

Specifications

Amperage 8A
Certifications/Compliance ETL, UL, CAN/CSA, CE
Temperature Range (Metric) 60°C
Voltage 115V
Display LCD
View More Specs
Requires: 
  • Freeze dry glassware if not bulk freeze drying
Products
Catalog Number Mfr. No. Weight (English) Weight (Metric) Price Quantity    

16-317-92

 
labconco™
LCN7806021
225 lb. 102kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

16-317-91

 
labconco™
LCN7806020
202 lb. 92kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Amperage 8A
Certifications/Compliance ETL, UL, CAN/CSA, CE
Temperature Range (Metric) 60°C
Voltage 115V
Display LCD
Electrical Requirements 115V 50/60Hz
For Use With (Equipment) 6, 12, or 18l freezone™ freeze-dry systems
Height (English) 27.1 in.
Height (Metric) 68.8cm
Hertz 50/60Hz
Interface RS232
Length (English) 19.5 in.
Length (English) Shelf 16.62 in.
Length (Metric) 49.5cm
Length (Metric) Shelf 42.2cm
No. of Shelves 3
Temperature Range (English) 140°F
Wattage 200w

Unique Shelf-heating System

  • Three 16.625L × 12.75 in.W (42.2 × 32.2cm) shelves, each with 210 sq. in. of space, accommodate bulk samples or batches of serum bottles, vials, or ampules (two additional shelves [order separately] can be added as needed) Microprocessor-controlled 200-watt heater on each shelf ensures efficient heating, up to +60°C, within 3°C of setpoint
  • Three sensor probes monitor sample or shelf temperature; can add two more sensors for a total of five (sensors sold separately)
User-friendly Operation
  • LCD display of setpoint and temperature of each shelf and indicates RUN (R) or STOP (S) modes
  • Vacuum release valve vents chamber to open door after drying
  • Rear-mounted RS-232 cable connection to IBM™-compatible computer; use connection to monitor parameters: shelf setpoint temperature, actual shelf temperature, run time, and operating status
Exterior Features
  • Clear 1 in. (2.5cm) thick acrylic viewing door
  • Cabinet is white powder-coated steel with a brushed stainless-steel front with blue accents
  • Integral support stand
  • Six-port manifold is included in 16-317-92; accommodates either 0.50 or 0.75 in. flask adapters to allow connection of flasks and freeze dry glassware so that samples attached to manifold and in Tray Dryer may be lyophilized simultaneously

Dimensions: 21.5L × 32W × 27 1/8 in.H (55 × 81 × 69cm)

ETL listed to UL standards (115V models), CAN/CSA (115V models), CE conformity (230V models).