missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ FlaskScrubber™ Pipet Inserts

Wash and dry pipets inside and out

Manufacturer:  Labconco™ LCN4494200

 View more versions of this product

Catalog No. 15-352-808F



Includes: PVC-coated holder

Description

Description

Wash and dry pipets inside and out

Specifications

Specifications

8 x 1 to 10mL pipet
Used for direct injection cleaning of pipets
1.25 in. mounting spindle, PVC coated holder
Pipet Insert
0.68kg
Each
FlaskScrubber™, FlaskScrubber™ Vantage series, SteamScrubber™ glassware washer
Type 304 Stainless Steel
1.5 lb.
Documents