Labconco™ Filters, Prefilters and Accessories
$353.36 - $1,104.05
Specifications
Filter Type HEPA
For Use With (Equipment) Precise™ HEPA filtered glove box, XPert™ weigh box
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Weight (Metric) Weight (English) Price Quantity  
Catalog Number Mfr. No. Weight (Metric) Weight (English) Price Quantity  
16-108-409 Labconco™
LCN5242400
2.3kg 5 lb. Each for $353.36
16-108-424 Labconco™
LCN5241700
0.7kg 1.5 lb. Each for $1,104.05
Specifications Description & Specifications
Specifications
HEPA
Precise™ HEPA filtered glove box, XPert™ weigh box