Labconco™ Fast-Freeze™ Flasks, Complete Assembly
Easy to handle and fast to load
Manufacturer:  Labconco™ LCN7540600
 View more versions of this product
Catalog No. 10-269-53
Requires : Adapters are required to attach flasks to freeze dry valves–40 and 80mL flasks use a 0.5 in. (1.3cm) adapter; larger sizes use a 0.75 in. (1.9cm).
Specifications Description & Specifications
Specifications
2.32 in.
59.2mm
5.7 in.
1.5 lb.
300mL
1.9cm
Compatible with all major brands of freeze-drying equipment
14.5cm
0.7kg
Borosilicate Glass