missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Fast-Freeze™ Flask Adapters

For connecting Fast-Freeze™ flasks to freeze-dryer valves

Manufacturer:  Labconco™ LCN7547600

 View more versions of this product

Catalog No. 10-269-61B


 

Specifications

Description

Adapter, Freeze Drying; Labconco; Fast-Freeze; Stainless Steel; 45 deg. Bend; 0.75 in.

  • 40 and 80mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.

Description & Specifications

Specifications

0.75 in.
For connecting fast freeze™ flasks to freeze-dryer valves
Stainless Steel
45° bend
0.2kg
1.9cm
Fast freeze™ flask
Freeze Drying Adapter
0.5 lb.