missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ CentriVap™ Rotors for Autosample Vials

$2,317.03 - $2,317.03

Specifications

For Use With (Equipment) CentriVap™ system
Product Type CentriVap™ Rotor
Material Thermoplastic
Shipping Weight (English) 3 lb.
Shipping Weight (Metric) 1.4kg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Holds Total Count Price Quantity  
Catalog Number Mfr. No. Holds Total Count Price Quantity  
16-317-70 Labconco™
LCN7824700
66 Autosampler vial 66.0 Each for $2,317.03
16-317-71 Labconco™
LCN7824702
56 Autosampler vial 56.0 Each for $2,317.03
Specifications Description & Specifications

Specifications

CentriVap™ system
Thermoplastic
1.4kg
1.4kg
CentriVap™ Rotor
3 lb.
3 lb.