missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Console FreeZone™ Freeze-Dry Systems, US Models: 18L

Available in several system configurations to meet sample load requirements

Manufacturer:  Labconco™ LCN7755041

 View more versions of this product

Catalog No. 10-272-01

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Requires : Drying chambers or other drying accessories, vacuum pumps and glassware. Require drying accessories.

Specifications Description & Specifications

Specifications

Console Freeze-Dry System
19 qt.
PTFE-coated Collector Chamber
7.5 in.
19 qt., 18L
30.5cm
208/230V
1.5hp
156kg
12.5 x 5.5 x 7.5 in. (30.5 x 14 x 19cm)
18L
≤10L
208/230V 60Hz
60Hz
345 lb.
19cm
14cm
5.5 in.
-50°C
12.5 in.