missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Console FreeZone™ Freeze-Dry Systems 208/230V: 6L

Freestanding models available in several system configurations and collector capacities to meet sample load requirements

Specifications

Capacity 6 qt., 6L
Dimensions (L x W x H) Exterior 28 x 31.875 x 36.25 in. (71 x 81 x 92cm)
Temperature Range (Metric) -50°C
Horsepower 3/4hp
Sublimation Rate <2L
View More Specs

Includes:
  • 8 ft. (2.4m) cord
  • Models wired include 3-wire cord with 20A NEMA 6-20P plug

Requires: Drying chambers or other drying accessories, vacuum pumps and glassware. All models require drying accessories.



Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Includes Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Includes Price Quantity  
10-271-42 Labconco™
LCN7753042
208/230V 60Hz Purge Valve N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-271-51 Labconco™
LCN7753544
230V 60Hz With Purge Valve, PTFE-coated Collector Chamber, Shell Freezer N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-274-68 Labconco™
LCN7758541
208/230V 60Hz Stoppering Tray Dryer and PTFE-coated collector chamber N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-274-67 Labconco™
LCN7758540
208/230V 60Hz Stoppering Tray Dryer N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-274-69 Labconco™
LCN7758542
208/230V 60Hz Purge Valve and Stoppering Tray Dryer N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-274-70 Labconco™
LCN7758544
230V 50Hz Stoppering Tray Dryer, PTFE-coated Collector Chamber, and Purge Valve N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Cabinets are white epoxy-coated steel with brushed stainless-steel front panels and are mounted on 3 in. (7.6cm) casters for mobility
  • Refrigeration system cools collector to -50°C (-58°F)
  • 4.5L model† comes with permanently installed 10-port stainless-steel drying chamber for bulk sample connection
  • Purge Valve option with switch prevents backstreaming of vacuum pump oil and minimizes chance for sample contamination of power if interrupted
  • Upright collector chamber simplifies defrosting; many models include built-in hot gas defrost to quickly melt any collected ice
  • Automatic one-button startup initiates condenser cooldown and vacuum pulldown
  • LCD indicates temperature in °C or °F; vacuum in mbar, Pa, and torr
  • LED vacuum and temperature “waves”, on all except 1L models, show relative system vacuum and collector temperature
  • LED alarm light, on all except 1L models, flashes for abnormal events, including power interruptions, improper power supply, when collector temperature rises above -40°C, and required vacuum pump service
  • Vacuum control valve, on all except 1L systems, maintains vacuum level

Connectivity

  • RS-232 port can be used to transmit data to a computer
  • Features
    • Easy to service with retractable drain hoses
    • Cabinets are white epoxy-coated steel with brushed stainless-steel front panels and blue accents
    • Console models have removable front panels and are roomy enough to accommodate vacuum pump
    • Benchtop models easily fit on laboratory benchtop to process light to moderate loads
    • PTFE-coated collector chamber and coil for processes involving corrosive compounds are available

    • Optional built-in shell freezer with stainless-steel bath, 12.5L × 5.5W × 7.5 in.H (30.5 × 14 × 19cm) for easier prefreezing
    • Available with optional built-in vacuum drying chamber (13 × 5 × 2.5 in. [33 × 12.9 × 6.4cm] that includes 50w heater with microprocessor control to +60°
    • Optional Stoppering Tray Dryer‡ accommodates serum bottles, vials, ampules, or bulk trays
    • Vacuum control valve maintains vacuum level

    Description & Specifications

    Specifications

    6 qt., 6L
    -50°C
    <2L
    28 x 31.875 x 36.25 in. (71 x 81 x 92cm)
    3/4hp