Please login to your online account to display your discounted pricing

Labconco™ Clear Stoppering Chambers


For use with 6L, 12L and 18L Freezone™ Dryers

Manufacturer: labconco™  LCN7868030

 View more versions of this product

Catalog No. 10-030-165

  • / Each
 

Description & Specifications

Specifications

Product Type Clear Stoppering Chamber
Diameter (English) 13.5 in.
Diameter (Metric) 34cm
Electrical Requirements 230V 50/60Hz
For Use With (Application) To provide economical means of stoppering serum bottles
For Use With (Equipment) For 6l, 12l and 18l freezone™ dryers
Height (English) 23 in.
Height (Metric) 58.4cm
Hertz 50/60Hz
Weight (English) 30 lb.
Weight (Metric) 13.6kg
Voltage 230V