missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Clear Drying Chambers for FreeZone™ Freeze-Dry Systems

For visible bulk drying

$2,487.07 - $4,822.19

Specifications

Color Clear
For Use With (Application) For visible bulk drying
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Height (English) Size Weight (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Height (English) Size Weight (English) Price Quantity  
10-030-154 Labconco™
LCN7442900
10.3 in. 13.1 in. Small 9.5 lb. Each for $2,487.07
10-030-155 Labconco™
LCN7443500
14.6 in. 12.1 in. Small 13.5 lb. Each for $3,462.69
10-271-1A Labconco™
LCN7867000
16.32 in. 13.37 in. Large 10 lb. Each for $3,446.69
10-030-156 Labconco™
LCN7444000
17.3 in. 15 in. Large 15 lb. Each for $4,822.19
Specifications

Description

  • Durable and corrosion-resistant
  • Chambers without valves include clear acrylic top and bottom lids, gaskets, vacuum break valve, and electrical pass-through for transformer cords
  • Chambers with valves include aluminum lid with eight valves that accommodate both 1/2 in. and 3/4 in. adapters for connecting flasks
  • Use with or without product shelves (shelves available separately, visit www.fishersci.com)

Description & Specifications

Specifications

Clear
For visible bulk drying