missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ CentriVap™ Solvent Systems

Combines multiple concentration components into one easy for ordering and just by adding vacuum pump it will be ready to concentrate samples.

Manufacturer:  Labconco™ 7995810

 View more versions of this product

Catalog No. 16-109-100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: Includes: Benchtop concentrator with glass lid, cold trap, solvent insert, clear canister, 12-13mm rotor, vacuum tubing and six clamps

Description

Description

Combines multiple concentration components into one easy for ordering and just by adding vacuum pump it will be ready to concentrate samples.

  • Benchtop style
  • Lid type: glass
  • Type/Application: solvents
  • Accessories (Required): Diaphragm or Rotary Vane vacuum pump

Specifications

Specifications

ETL
Benchtop concentrator with Glass lid, cold trap, solvent insert, clear canister, 12 to 13mm rotor, vacuum tubing and 6 clamp
115 V 60 Hz
Domestic - North America
115 V
60 Hz
8.5 A
SDS
Documents
Product Certifications
Promotions

Promotions