Labconco™ Carbon Exhaust Flow Filters for Trace Organics
Trace Odor Carbon Filters for Labconco Purifier and XPert Enclosures.
Supplier: Labconco™ 3937402
Description
Trace Odor Carbon Filters for easy installation atop the XPert and RXPert Filtered Balance Systems and Purifier Class I and HEPA Filtered Enclosures. Filters trap non-volatile nuisance odors and vapors from samples or minute quantities of chemicals. Whether or not a Trace Odor Carbon Filter is appropriate for your application requires consideration of factors such as the time-weighted exposure limits of organic solvents, formaldehyde, ammonia-amines or acid-sulfur.Specifications
Carbon | |
1 in. | |
8.6 Kg | |
18 in. | |
2.5 cm | |
45.7 cm | |
106.7 cm | |
Ammonia, Amines |
4 ft. Purifier™ class I enclosures, Purifier™ HEPA filtered enclosures, XPert™ filtered balance system | |
19 lb. | |
40 to 50 in. | |
42 in. | |
40 to 50 x 42 x 1 in. (101.6 to 127 x 106.7 x 2.5 cm) | |
101.6 to 127 cm | |
Filter |