Labconco™ Canopy Connections
For Purifier™ Class I and HEPA-filtered Enclosures and XPert™ Filtered Balance Systems
Supplier:  Labconco™ 3924401
 View more versions of this product
Catalog No. 10-304-151
Description
Description
Allow Purifier Class I and HEPA-filtered Enclosures and XPert Filtered Balance Systems to be thimble-ducted to the outside for additional protection.
  • Canopy's air gap provides means of balancing air by allowing room air to enter ductwork and mix with filtered air being exhausted from enclosure
  • Outlet: 6.06 in. I.D. and 6.19 in. O.D. and accepts 6 in. nominal diameter ductwork
Specifications
Specifications
Purifier Class I or HEPA Filtered Enclosure (36 in.
8.2 Kg
15.24 cm
18 lb.
6 in.
Balancing Air by Allowing Room Air to Enter the Ductwork