Please login to your online account to display your discounted pricing

Labconco™ Benchtop Fume Adsorbers-2

Designed for light-duty applications involving organic solvents, formaldehyde, ammonia or low molecular weight amines

$7,105.25 - $7,105.25

Products
Catalog Number Mfr. No. Price Quantity    

10-090-102

 
labconco™
LCN3955400
Each for $7,105.25
Description & Specifications

  • Two built-in motorized impellers to provide fast velocity up to 35fp
  • Glacier white, dry powder, epoxy-coated steel housing with perforated baffle for uniform airflow
  • Clear, one-piece, 3/8 in. thick, shatter-resistant acrylic canopy
  • Quiet operation at 58-63 dBA
  • Pre-wired with rear-mounted auxiliary electrical receptacle and top-mounted blower switch with circuit breaker
  • Rear-mounted air sampling port
  • Exterior dimensions: 15.4 x 23.4 x 60 in. (H x D x W) (39.1 x 59.4 x 152.4cm)
  • Interior dimensions: 13.5 x 14.5 in. (H x D) (34.2 x 36.8cm)

Model 3955402 (10-090-124):

  • Features two each replaceable 8.8 lb impregnated carbon filter treated to adsorb ammonia and low molecular weight amine vapors. Adsorption rate 10% (0.88 lb)
  • Operates on 115V 60Hz, 2 amps
  • 6.5 ft IEC three-wire cord with plug
Models 3955420, 3955421 and 3955422 (10-090-125, 10-090-126 and 10-090-127)
  • Operation on 240V 50Hz, 1 amp
  • 9 ft IEC three-wire cord (electrical plug not included)

All models conform to the following regulations and standards:

  • UL 3101-1/61010-1 (115V models)
  • CAN/CSA C22.2 No. 1010.0 (115V models)
  • CE Conformity Marking (230V models)