missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Benchtop FreeZone™ Freeze-Dry Systems, US Models: 4.5L

With a small footprint and four rubber feet, these smaller capacity models fit easily on benchtop and are ideal for light and moderate loads

Manufacturer:  Labconco™ LCN7750020

 View more versions of this product

Catalog No. 1027110

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Requires : Drying chambers or other drying accessories, vacuum pumps and glassware. Come with permanently installed 10-port stainless-steel drying chamber for bulk sample connection.

Specifications Description & Specifications

Specifications

4.5L
115V 60Hz
Stainless-steel coil and collector chamber
>2L
22.5 in.
115V
18.5 in.
60Hz
Benchtop Freeze-Dry System
18.6 in.
-50°C