missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Benchtop FreeZone™ Freeze-Dry Systems, 208/230V 60Hz Models: 4.5L

With small footprint and four feet, this smaller capacity model fits easily on benchtop and is ideal for light and moderate loads

Specifications

Capacity (Metric) 4.5L
Temperature Range (Metric) -50°C
Electrical Requirements 208/230V 60Hz
Sublimation Rate >2L
View More Specs

Includes: 8 ft. (2.4m) cord, 3-wire cord with 20A NEMA 6-20P plug

Requires: Required Accessories

  • Drying chambers or other drying accessories, vacuum pumps and glassware
  • Come with permanently installed 10-port stainless-steel drying chamber for bulk sample connection



Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
10-271-15 Labconco™
LCN7750041
PTFE-coated collector chamber N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-271-14 Labconco™
LCN7750040
Stainless-steel coil and collector chamber N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Refrigeration system cools collector to -50°C (-58°F)
  • Has stainless-steel drying chamber with 10 valves to connect serum bottles, ampules or freeze-dry flasks with 1/2 to 3/4 in. adapters (available separately)
  • Upright collector chamber simplifies defrosting; many models include built-in hot gas defrost to quickly melt any collected ice
  • Automatic one-button startup initiates condenser cooldown and vacuum pulldown
  • LCD indicates temperature in °C or °F; vacuum in mbar, Pa, and torr
  • LED vacuum and temperature “waves” show relative system vacuum and collector temperature
  • LED alarm light flashes for abnormal events, including power interruptions, improper power supply, when collector temperature rises above -40°C, and required vacuum pump service
  • Vacuum control valve maintains vacuum level

Connectivity

  • RS-232 port can be used to transmit data to a computer
  • Features
    • Easy to service with retractable drain hoses
    • Cabinets are white epoxy-coated steel with brushed stainless-steel front panels and blue accents
    • Console models have removable front panels and are roomy enough to accommodate vacuum pump
    • Benchtop models easily fit on laboratory benchtop to process light to moderate loads
    • PTFE-coated collector chamber and coil for processes involving corrosive compounds are available

    Order Info

    Contact your Fisher Scientific Customer Service Representative for pump compatibility.

    †Overall dimensions include Drying Chamber.

    Description & Specifications

    Specifications

    4.5L
    -50°C
    208/230V 60Hz
    >2L