missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinets, 6 ft. W, 8 in. Opening
Features ergonomic Inclination™ Technology and MyLogic™ Operating System
Manufacturer: Labconco™ LCN304681010
Description
The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.
- Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
- Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
- Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes
The new C1 designation is pending NSF approval
Specifications
32.6 in. | |
230V, 50/60Hz, 6A, Domestic | |
UV light, service fixture and Vacu-Pass Portal | |
Safety Cabinet | |
900 lb. | |
78.2 in. | |
NSF International-Listed, ETL, Modified ASHRAE 110 | |
175.2cm | |
20.3cm |
82.8cm | |
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards | |
Fluorescent | |
8 in. | |
408.2kg | |
198.6cm | |
69 in. | |
North America |