missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinets, 6 ft. W, 8 in. Opening

Features ergonomic Inclination™ Technology and MyLogic™ Operating System

Specifications

Product Type Safety Cabinet
Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110
For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Sash Opening (Metric) 20.3cm
Width (Metric) 198.6cm
View More Specs

Products 8
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Height (Metric) Price Quantity  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Height (Metric) Price Quantity  
30-468-0000
View Documents
Labconco™
LCN304680000
North America 115V, 60Hz, 12A, Domestic N/A 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-0100
View Documents
Labconco™
LCN304680100
North America 115V, 60Hz, 12A, Domestic Base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-1000
View Documents
Labconco™
LCN304681000
North America 115V, 60Hz, 12A, Domestic UV light, service fixture and Vacu-Pass Portal 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-1100
View Documents
Labconco™
LCN304681100
North America 115V, 60Hz, 12A, Domestic UV light, service fixture, Vacu-Pass Portal, Base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-0010
View Documents
Labconco™
LCN304680010
North America 230V, 50/60Hz, 6A, Domestic N/A 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-0110
View Documents
Labconco™
LCN304680110
North America 230V, 50/60Hz, 6A, Domestic Base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-1010
View Documents
Labconco™
LCN304681010
North America 230V, 50/60Hz, 6A, Domestic UV light, service fixture and Vacu-Pass Portal 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-468-1110
View Documents
Labconco™
LCN304681110
North America 230V, 50/60Hz, 6A, Domestic UV light, service fixture, Vacu-Pass Portal, Base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.

 

  • Safe and easy to operate than traditional biosafety cabinets due to efficient, well-defined airflow pattern over its work surface
  • Chem-Zone™(patent pending) increases chemical safety by creating large, clearly defined area within the enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, w/option of venting away hazardous fumes

 

The new C1 designation is pending NSF approval

Specifications

Specifications

Safety Cabinet
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
198.6cm
Fluorescent
NSF International-Listed, ETL, Modified ASHRAE 110
20.3cm
82.8cm
SDS