missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinets, 4 ft. W, 8 in. Opening

Features ergonomic Inclination™ Technology and MyLogic™ Operating System

Manufacturer:  Labconco™ LCN304481100

 View more versions of this product

Catalog No. 30-448-1100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.

 

  • Efficient airflow pattern over work surface makes cabinet safe and easy to operate than traditional biosafety cabinets
  • Chem-Zone™ design maximizes chemical safety by creating large, clearly defined area within enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols (such as agents that require Biosafety Level 1, 2 or 3 containment, with the option of venting away hazardous fumes)

 

The new C1 designation is pending NSF approval

Specifications

Specifications

32 in.
115V, 60Hz, 12A, Domestic
UV light, service fixture and base stand
Safety Cabinet
721 lb.
54.2 in.
NSF International-Listed, ETL, Modified ASHRAE 110
245.1cm
20.3cm
81.2cm
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Fluorescent
8 in.
327kg
137.6cm
96.5 in.
North America
SDS
Documents