missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Axiom™ Class II, Type C1 Biosafety Cabinets, 4 ft. W, 8 in. Opening

Features ergonomic Inclination™ Technology and MyLogic™ Operating System

Specifications

Product Type Safety Cabinet
Certifications/Compliance NSF International-Listed, ETL, Modified ASHRAE 110
For Use With (Application) Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
Sash Opening (Metric) 20.3cm
Width (Metric) 137.6cm
View More Specs

Products 9
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Height (Metric) Price Quantity  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Height (Metric) Price Quantity  
30-448-0000
View Documents
Labconco™
LCN304480000
North America 115V, 60Hz, 12A, Domestic N/A 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-0100
View Documents
Labconco™
LCN304480100
North America 115V, 60Hz, 12A, Domestic Base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-1000
View Documents
Labconco™
LCN304481000
North America 115V, 60Hz, 12A, Domestic UV light, service fixture and Vacu-Pass Portal 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-1100
View Documents
Labconco™
LCN304481100
North America 115V, 60Hz, 12A, Domestic UV light, service fixture and base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-0010
View Documents
Labconco™
LCN304480010
North America 230V, 50/60Hz, 6A, Domestic N/A 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-0110
View Documents
Labconco™
LCN304480110
North America 230V, 50/60Hz, 6A, Domestic Base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-1010
View Documents
Labconco™
LCN304481010
North America 230V, 50/60Hz, 6A, Domestic UV light, service fixture and Vacu-Pass Portal 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-1110
View Documents
Labconco™
LCN304481110
North America 230V, 50/60Hz, 6A, Domestic UV light, service fixture, Vacu-Pass Portal and base stand 245.1cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
30-448-0020
View Documents
Labconco™
LCN304480020
Japan 100V, 50/60Hz, 12A, International N/A 175.2cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The Purifier Axiom Class II, Type C1 is first biological safety cabinet that can be used in recirculating Type A-mode for standard microbiological work or can be connected to exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides.

 

  • Efficient airflow pattern over work surface makes cabinet safe and easy to operate than traditional biosafety cabinets
  • Chem-Zone™ design maximizes chemical safety by creating large, clearly defined area within enclosure
  • Provides protection for personnel, product and laboratory environment by filtering hazardous particulates and aerosols (such as agents that require Biosafety Level 1, 2 or 3 containment, with the option of venting away hazardous fumes)

 

The new C1 designation is pending NSF approval

Specifications

Specifications

Safety Cabinet
Applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards
137.6cm
Fluorescent
NSF International-Listed, ETL, Modified ASHRAE 110
20.3cm
81.2cm
SDS