missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer (Ready-To-Use)

Take advantage of a one-step polymer system providing increased sensitivity, time savings and detection simplicity with the Thermo Scientific™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer (Ready-To-Use).

$1,520.49 - $2,500.35

Specifications

Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme Peroxidase
Storage Requirements Store at 2° to 8°C. Each component is stable for 18 months.
Substrate None provided
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
TL060HLJ Thermo Scientific™
TL060HLJ
60mL Each for $1,520.49
TL125HLJ Thermo Scientific™
TL125HLJ
125mL Each for $2,500.35
Specifications

Description

Take advantage of a one-step polymer system providing increased sensitivity, time savings and detection simplicity with the Thermo Scientific™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer (Ready-To-Use).

UltraVision ONE is a robust ONE-step polymer system that provides increased sensitivity, time savings and detection simplicity. The UltraVision ONE HRP polymer is an innovative, patented technology. It consists of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. UltraVision ONE HRP polymer allows the use of less antibody to obtain better signal-to-noise ratios. This system is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods. For optimal interpretation of results, appropriate positive and negative controls must be included.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes:

TL-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE HRP Polymer (TL-060-PHJ)

TL-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE HRP Polymer (TL-125-PHJ)

Note:

UltraVision ONE HRP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to horseradish peroxidase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.

Description & Specifications

Specifications

None provided
Peroxidase
None provided
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Store at 2° to 8°C. Each component is stable for 18 months.