Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ UltraVision™ LP Detection System: AP Polymer (Ready-To-Use)

Take advantage of a senstive detection system based on biotin-free, polymer technology with the Thermo Scientific™ Lab Vision™ UltraVision™ LP Detection System: AP Polymer.

$1,842.41 - $3,044.16

Specifications

Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme Alkaline Phosphatase
Substrate None provided
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

TL060AL

 
thermo scientific™
TL060AL
60mL Each for $1,842.41

TL125AL

 
thermo scientific™
TL125AL
125mL Each for $3,044.16
Description & Specifications

Specifications

Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme Alkaline Phosphatase
Substrate None provided

Take advantage of a senstive detection system based on biotin-free, polymer technology with the Thermo Scientific™ Lab Vision™ UltraVision™ LP Detection System: AP Polymer.

UltraVision LP is the latest technology in polymeric labeling. Polymer detection methods have been shown to provide increased sensitivity and detection simplicity. This second-generation polymer system is composed of smaller polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. With UltraVision LP, you use less antibody and obtain better signal-to-noise ratios. UltraVision LP is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods.

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Alkaline Phosphatase

  • Chromogen/Substrate: None provided

Includes:

TL060AL: Ultra V Block (Mfr. No. TA-060-UB), Primary Antibody Enhancer (Mfr. No. TL-060-PB), AP Polymer (Mfr. No. TL-060-AP)

TL125AL: Ultra V Block (Mfr. No. TA-125-UB), Primary Antibody Enhancer (Mfr. No. TL-125-PB), AP Polymer (Mfr. No. TL-125-AP)

Note:

UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.