missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynalon™ Lab Supply Container

Lightweight container has a wide range of uses in the lab.

Manufacturer:  Dynalon™ DAL410545

 View more versions of this product

Catalog No. S47884A




Description

Description

Tote tray of molded, seamless polypropylene. This sterilizing unit has countless uses in science laboratories. • Autoclavable • Lightweight • Tapered for nesting
Specifications

Specifications

13 cm
Polypropylene
26 cm
5.125 in.
CS/6, PK/1
19 x 11 x 5 in. (48 x 26 x 13cm)
Autoclavable
Natural
48 cm
Tote Box
19 x 11 x 5 in. (48 x 26 x 13 cm)
19 in.
Rectangular
10.25 in.
16 L
SDS
Documents
Product Certifications
Promotions

Promotions