missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dynalon™ Lab Supply Container
Lightweight container has a wide range of uses in the lab.
Manufacturer: Dynalon™ DAL410535
Description
Tote tray of molded, seamless polypropylene. This sterilizing unit has countless uses in science laboratories. • Autoclavable • Lightweight • Tapered for nestingSpecifications
13.3 cm | |
Polypropylene | |
20 cm | |
5.2 in. | |
CS/6, PK/1 | |
7.8 in. | |
7.8L |
31 cm | |
Tote Box | |
12.2 x 7.8 x 5.2 in. (31 x 20 x 13.3 cm) | |
12.2 in. | |
12.2 x 7.8 x 5.2 in. (31 x 20 x 13.3cm) | |
Autoclavable | |
Natural |